siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 11 | 1737 | ||
AF402612.1 | - | 10 | 1634 | |||
AF402613.1 | - | 11 | 1784 | |||
AK028302.1 | - | 8 | 1186 | |||
AK034504.1 | - | 10 | 1727 | |||
AK134961.1 | - | 11 | 1800 | |||
AK149626.1 | - | 11 | 1788 | |||
BC129902.1 | - | 11 | 1824 | 40111637 | ||
BC129903.1 | - | 11 | 1825 | 40111639 | ||
BC145523.1 | - | 11 | 1878 |
