siRNA Details
Gene Symbol: Xdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011723.3 | NP_035853.2 | 23 | 2569 | ||
GenBank mRNA | AK034256.1 | - | 23 | 2517 | ||
AK090058.1 | - | 14 | 1710 | |||
AK134156.1 | - | 9 | 1054 | |||
AK143412.1 | - | 23 | 2518 | |||
AK144700.1 | - | 23 | 2521 | |||
AK160010.1 | - | 20 | 2268 | |||
AK161529.1 | - | 23 | 2520 | |||
AK171642.1 | - | 4 | 411 | |||
BC032077.1 | - | 7 | 774 | |||
BC065699.1 | - | 14 | 1706 | |||
BC141183.1 | - | 23 | 2544 | 9055795 | ||
X62932.1 | - | 23 | 2503 |
