siRNA Details
Gene Symbol: Xdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011723.3 | NP_035853.2 | 3, 4 | 266 | ||
GenBank mRNA | AK034256.1 | - | 3, 4 | 216 | ||
AK143412.1 | - | 3, 4 | 215 | |||
AK144700.1 | - | 3, 4 | 218 | |||
AK161529.1 | - | 3, 4 | 217 | |||
AK164764.1 | - | 3, 4 | 209 | |||
BC141183.1 | - | 3, 4 | 241 | 9055795 | ||
X62932.1 | - | 3, 4 | 200 |
