siRNA Details
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145948.1 | NP_001139420.1 | 15 | 1353 | ||
NM_153392.2 | NP_700441.2 | 15 | 1362 | |||
GenBank mRNA | AK048659.1 | - | 15 | 1363 | ||
AK076641.1 | - | 15 | 1316 | |||
AK076720.1 | - | 15 | 1355 | |||
AK076772.1 | - | 15 | 1317 | |||
AK172951.1 | - | 15 | 1285 | |||
BC023883.1 | - | 15 | 1361 | 5150841 | ||
BC026660.1 | - | 15 | 1303 | 4988900 |
