siRNA Details
Gene Symbol: Paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198422.2 | NP_940814.1 | 4 | 796 | ||
GenBank mRNA | AK149213.1 | - | 4 | 775 | ||
AK172689.1 | - | 4 | 752 | |||
AY424292.1 | - | 4 | 796 | |||
BC120853.1 | - | 4 | 781 | 40129776 | ||
BC120855.1 | - | 4 | 782 | 40129778 |
