siRNA Details
Gene Symbol: Paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198422.2 | NP_940814.1 | 3 | 587 | ||
GenBank mRNA | AK149213.1 | - | 3 | 566 | ||
AK172689.1 | - | 3 | 542 | |||
AY424292.1 | - | 3 | 587 | |||
BC050248.1 | - | 3 | 587 | |||
BC051173.1 | - | 3 | 587 | |||
BC120853.1 | - | 3 | 572 | 40129776 | ||
BC120855.1 | - | 3 | 573 | 40129778 |
