siRNA Details
Gene Symbol: Arglu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176849.3 | NP_789819.2 | 2 | 742 | ||
GenBank mRNA | AK145733.1 | - | 2 | 545 | ||
BC029219.1 | - | 1 | 145 | |||
BC046971.1 | - | 3 | 672 | 6468505 | ||
BU514203.1 | - | 3 | 389 | |||
BY756892.1 | - | 2 | 516 |
