siRNA Details
Gene Symbol: Tmem205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253867.1 | NP_001240796.1 | 3, 4 | 439 | ||
NM_001253868.1 | NP_001240797.1 | 3, 4 | 512 | |||
NM_001253869.1 | NP_001240798.1 | 3, 4 | 371 | |||
NM_001253870.1 | NP_001240799.1 | 3, 4 | 499 | |||
NM_178577.5 | NP_848692.2 | 3, 4 | 507 | |||
GenBank mRNA | AB030201.1 | - | 3, 4 | 342 | ||
AI573565.1 | - | 3, 4 | 323 | |||
BC010787.1 | - | 3, 4 | 434 | 4211629 | ||
CA326029.1 | - | 3, 4 | 371 | |||
CD775257.1 | - | 1, 2 | 365 | |||
CN840781.1 | - | 3, 4 | 427 |
