siRNA Details
Gene Symbol: 4833427G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177702.3 | NP_808370.1 | 4, 5 | 507 | ||
GenBank mRNA | AK014780.1 | - | 4, 5 | 396 | ||
AK132837.1 | - | 4, 5 | 470 | |||
BC048587.1 | - | 4, 5 | 507 | 6707436 |
