siRNA Details
Gene Symbol: 1700071K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033765.2 | NP_001028937.1 | 1 | 83 | ||
GenBank mRNA | AK131752.1 | - | 1 | 84 | ||
BC139179.1 | - | 1 | 49 | 8862201 | ||
BC139181.1 | - | 1 | 49 | 8862203 |

More Information
Other Species Targeted

