siRNA Details
Gene Symbol: 1700084C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033185.2 | NP_001028357.1 | 2 | 138 | ||
GenBank mRNA | AK141447.1 | - | 2 | 139 | ||
BC150846.1 | - | 1 | 63 | 9087757 | ||
BC172001.1 | - | 1 | 63 | 9054287 |
