siRNA Details
Gene Symbol: Dclk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172928.5 | NP_766516.2 | 2 | 1903 | ||
GenBank mRNA | AK050312.1 | - | 2 | 1871 | ||
AK135010.1 | - | 2 | 1892 | |||
BC056929.1 | - | 1 | 1571 | 6816292 | ||
DQ286388.1 | - | 2 | 1872 |
