siRNA Details
Gene Symbol: Olfr247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146269.2 | NP_666381.2 | 36 |
Gene Symbol: Olfr819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165944.1 | NP_001159416.1 | 1 | 54 |
