siRNA Details
Gene Symbol: Olfr899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146479.1 | NP_666690.1 | 1 | 166 | ||
GenBank mRNA | BC104124.1 | - | 1 | 166 | 40044544 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146479.1 | NP_666690.1 | 1 | 166 | ||
GenBank mRNA | BC104124.1 | - | 1 | 166 | 40044544 |