siRNA Details
Gene Symbol: Olfr197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146484.1 | NP_666695.1 | 1 | 476 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146484.1 | NP_666695.1 | 1 | 476 |