siRNA Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008927.3 | NP_032953.1 | 2 | 562 | ||
GenBank mRNA | AB041567.1 | - | 2 | 542 | ||
AK146233.1 | - | 2 | 552 | |||
AK146931.1 | - | 2 | 555 | |||
AK154793.1 | - | 2 | 589 | |||
AK165664.1 | - | 1 | 96 | |||
AK165894.1 | - | 2 | 563 | |||
AK167336.1 | - | 2 | 529 | |||
BC051137.1 | - | 2 | 381 | |||
BC054754.1 | - | 2 | 341 | 6411664 | ||
L02526.1 | - | 2 | 289 |
