siRNA Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008927.3 | NP_032953.1 | 7, 8 | 1204 | ||
GenBank mRNA | AK146233.1 | - | 7, 8 | 1194 | ||
AK146931.1 | - | 7, 8 | 1197 | |||
AK154793.1 | - | 7, 8 | 1231 | |||
AK165664.1 | - | 6, 7 | 738 | |||
AK165894.1 | - | 7, 8 | 1205 | |||
AK167336.1 | - | 7, 8 | 1171 | |||
AK192210.1 | - | 2, 3 | 256 | |||
AK214848.1 | - | 256 | ||||
BC051137.1 | - | 7, 8 | 1023 | |||
BC054754.1 | - | 7, 8 | 983 | 6411664 | ||
L02526.1 | - | 7, 8 | 931 |
