siRNA Details
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183392.2 | NP_899248.1 | 10 | 1373 | ||
GenBank mRNA | AK088855.1 | - | 10 | 1298 | ||
AK133841.1 | - | 10 | 1371 | |||
AK150182.1 | - | 10 | 1298 | |||
AK150841.1 | - | 10 | 1260 | |||
AK151896.1 | - | 10 | 1260 | |||
AK152136.1 | - | 10 | 1260 | |||
AK162037.1 | - | 10 | 1305 | |||
BC044303.1 | - | 10 | 1298 | |||
BC068114.1 | - | 10 | 1258 | 5707888 |
