siRNA Details
Gene Symbol: H2afy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159513.1 | NP_001152985.1 | 9 | 1318 | ||
NM_001159514.1 | NP_001152986.1 | 9 | 1312 | |||
NM_001159515.1 | NP_001152987.1 | 9 | 1309 | |||
NM_012015.2 | NP_036145.1 | 9 | 1321 | |||
GenBank mRNA | AB071988.2 | - | 9 | 1257 | ||
AF123312.1 | - | 2 | 272 | |||
AF171080.1 | - | 9 | 1299 | |||
AK003531.1 | - | 1 | 89 | |||
AK179948.1 | - | 1 | 107 | |||
AK214939.1 | - | 1 | 107 | |||
BC006955.1 | - | 9 | 1256 | 2811919 | ||
CB574813.1 | - | 6 | 630 | |||
CV561613.1 | - | 6 | 645 |
