siRNA Details
Gene Symbol: H2afy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159513.1 | NP_001152985.1 | 3 | 485 | ||
NM_001159514.1 | NP_001152986.1 | 3 | 485 | |||
NM_001159515.1 | NP_001152987.1 | 3 | 485 | |||
NM_012015.2 | NP_036145.1 | 3 | 485 | |||
GenBank mRNA | AB071988.2 | - | 3 | 421 | ||
AF171080.1 | - | 3 | 463 | |||
AK039274.1 | - | 3 | 466 | |||
AK143498.1 | - | 3 | 376 | |||
BB633156.1 | - | 3 | 466 | |||
BC006955.1 | - | 3 | 429 | 2811919 |
