siRNA Details
Gene Symbol: Bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012060.4 | NP_036190.2 | 4 | 403 | ||
GenBank mRNA | AK008043.1 | - | 4 | 403 | ||
AK011500.1 | - | 4 | 404 | |||
AK151641.1 | - | 4 | 404 | |||
BC002106.1 | - | 4 | 371 | 3486784 | ||
X81816.1 | - | 3 | 303 |
