siRNA Details
Gene Symbol: Srr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163311.1 | NP_001156783.1 | 4 | 532 | ||
NM_013761.4 | NP_038789.1 | 5 | 758 | |||
GenBank mRNA | AF148321.1 | - | 4 | 413 | ||
AK017235.1 | - | 5 | 637 | |||
AK028034.1 | - | 4 | 415 | |||
AK031687.1 | - | 5 | 630 | |||
AK043738.1 | - | 5 | 664 | |||
AK043819.1 | - | 4 | 438 | |||
AK080830.1 | - | 5 | 638 | |||
AK157122.1 | - | 4 | 683 | |||
AK170096.1 | - | 5 | 644 | |||
AK170471.1 | - | 5 | 644 | |||
BC011164.1 | - | 5 | 635 | 4195695 |
