siRNA Details
Gene Symbol: Naglu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013792.2 | NP_038820.2 | 6 | 1426 | ||
GenBank mRNA | AK143546.1 | - | 6 | 1428 | ||
AK158774.1 | - | 4 | 2050 | |||
AK164125.1 | - | 6 | 1405 | |||
AK170758.1 | - | 6 | 1409 | |||
AK185044.1 | - | 1 | 378 | |||
AK185937.1 | - | 1 | 258 | |||
AK189169.1 | - | 2 | 418 | |||
AK215965.1 | - | 1 | 133 | |||
BC055733.1 | - | 6 | 1400 | 6411530 | ||
U85247.1 | - | 6 | 1395 |
