siRNA Details
Gene Symbol: Fam21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026585.3 | NP_080861.2 | 23 | 2415 | ||
GenBank mRNA | AK038318.1 | - | 23 | 2393 | ||
AK043764.1 | - | 12 | 1260 | |||
AK122320.1 | - | 22 | 2421 | |||
AK145716.1 | - | 23 | 2416 | |||
AK161181.1 | - | 23 | 2365 | |||
AK163765.1 | - | 22 | 2403 | |||
AK168388.1 | - | 23 | 2404 | |||
AK190151.1 | - | 3 | 223 | |||
BC032177.1 | - | 21 | 2641 | |||
BC040393.1 | - | 23 | 2339 | |||
BC049979.1 | - | 23 | 2391 | |||
BC056942.1 | - | 23 | 2342 | 6827177 |
