siRNA Details
Gene Symbol: Olfr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019474.2 | NP_062347.2 | 1 | 313 | ||
GenBank mRNA | BC101957.1 | - | 1 | 313 | 40044987 | |
BC101961.1 | - | 1 | 313 | 40044981 | ||
BC103551.1 | - | 1 | 313 | 40044986 | ||
BC104727.1 | - | 1 | 313 | 40044983 |
