siRNA Details
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159284.1 | NP_001152756.1 | 10, 11 | 1232 | ||
NM_001284427.1 | NP_001271356.1 | 9, 10 | 1181 | |||
NM_001284428.1 | NP_001271357.1 | 10, 11 | 1289 | |||
NM_001284429.1 | NP_001271358.1 | 10, 11 | 1234 | |||
NM_013870.3 | NP_038898.2 | 9, 10 | 1181 | |||
GenBank mRNA | AF064236.1 | - | 9, 10 | 1128 | ||
AJ010305.1 | - | 9, 10 | 1128 | |||
AK030932.1 | - | 9, 10 | 1182 | |||
AK085412.1 | - | 10, 11 | 1196 | |||
AK138898.1 | - | 10, 11 | 1246 | |||
AK166385.1 | - | 10, 11 | 1249 | |||
AK170516.1 | - | 9, 10 | 1141 | |||
BC010599.1 | - | 9, 10 | 1122 | 3709078 | ||
BC066192.1 | - | 10, 11 | 1110 | 30433616 | ||
BC069836.1 | - | 10, 11 | 1110 | 30456568 | ||
BC069839.1 | - | 10, 11 | 1110 | 30431675 |
