siRNA Details
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 6 | 970 | ||
GenBank mRNA | AJ299404.1 | - | 6 | 938 | ||
AK007814.1 | - | 6 | 959 | |||
AK009693.1 | - | 6 | 988 | |||
AK017908.1 | - | 6 | 915 | |||
AK151048.1 | - | 6 | 956 | |||
AK151390.1 | - | 7 | 1007 | |||
AK152243.1 | - | 6 | 935 | |||
AK152358.1 | - | 6 | 971 | |||
AK152484.1 | - | 6 | 971 | |||
AK153391.1 | - | 6 | 956 | |||
AK170296.1 | - | 6 | 965 | |||
AK172355.1 | - | 6 | 945 | |||
BC025525.1 | - | 6 | 954 | |||
BC052851.1 | - | 6 | 1259 | |||
BC071216.1 | - | 6 | 951 | 30439199 |
