siRNA Details
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 3, 4 | 626 | ||
GenBank mRNA | AJ299404.1 | - | 3, 4 | 594 | ||
AK007814.1 | - | 3, 4 | 615 | |||
AK009693.1 | - | 3, 4 | 644 | |||
AK017908.1 | - | 3, 4 | 571 | |||
AK152243.1 | - | 3, 4 | 591 | |||
AK152358.1 | - | 3, 4 | 627 | |||
AK152484.1 | - | 3, 4 | 627 | |||
AK170296.1 | - | 3, 4 | 621 | |||
AK172355.1 | - | 3, 4 | 601 | |||
BC025525.1 | - | 3, 4 | 611 | |||
BC052851.1 | - | 3, 4 | 915 | |||
BC071216.1 | - | 3, 4 | 607 | 30439199 |
