siRNA Details
Gene Symbol: Tmem243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081029.1 | NP_001074498.1 | 4, 5 | 456 | ||
GenBank mRNA | AK076719.1 | - | 3, 4 | 318 | ||
BC147127.1 | - | 3, 4 | 283 | 9056179 | ||
BC147128.1 | - | 3, 4 | 283 | 9056187 | ||
BC147812.1 | - | 3, 4 | 283 | 9087529 | ||
BC147813.1 | - | 3, 4 | 283 | 9087530 |
