siRNA Details
Gene Symbol: Dlx6os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032180.1 | - | 2 | 154 | ||
AK032537.1 | - | 2 | 254 | |||
AK038694.1 | - | 2 | 96 | |||
AK044034.1 | - | 2 | 113 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dlx6os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032180.1 | - | 2 | 154 | ||
AK032537.1 | - | 2 | 254 | |||
AK038694.1 | - | 2 | 96 | |||
AK044034.1 | - | 2 | 113 |