siRNA Details
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177081.3 | NP_796055.1 | 7 | 809 | ||
GenBank mRNA | AK041415.1 | - | 4 | 2850 | ||
AK049175.1 | - | 6 | 660 | |||
AK083305.1 | - | 7 | 809 | |||
BC098475.1 | - | 7 | 863 | 30044369 | ||
BC138194.1 | - | 6 | 745 | 8861213 |
