siRNA Details
Gene Symbol: Susd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163288.2 | NP_001156760.2 | 12 | 1916 | ||
GenBank mRNA | AK041075.1 | - | 3 | 194 | ||
BC125408.1 | - | 2 | 74 | |||
BC125410.1 | - | 2 | 74 | |||
BC157977.1 | - | 12 | 1841 | 9088138 | ||
BC158124.1 | - | 12 | 1893 | 9007393 |
