siRNA Details
Gene Symbol: Tcrb-J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017898.1 | - | 4 | 513 | ||
M11456.1 | - | 2 | 334 | |||
M13676.1 | - | 2 | 307 | |||
M16679.1 | - | 2 | 304 | |||
M20877.1 | - | 1 | 256 | |||
M20878.1 | - | 1 | 256 | |||
U21402.1 | - | 58 | ||||
U26785.1 | - | 64 | ||||
U26791.1 | - | 1 | 49 | |||
U26795.1 | - | 1 | 49 | |||
U26796.1 | - | 1 | 49 | |||
U46842.1 | - | 2 | 307 | |||
X00438.1 | - | 2 | 329 | |||
X02778.1 | - | 2 | 283 | |||
X02779.1 | - | 2 | 307 |
