siRNA Details
Gene Symbol: Tcrb-J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017898.1 | - | 4 | 432 | ||
M11456.1 | - | 2 | 253 | |||
M13676.1 | - | 2 | 226 | |||
M16679.1 | - | 2 | 223 | |||
M20877.1 | - | 1 | 175 | |||
M20878.1 | - | 1 | 175 | |||
U46842.1 | - | 2 | 226 | |||
X00438.1 | - | 2 | 248 | |||
X02778.1 | - | 2 | 202 | |||
X02779.1 | - | 2 | 226 |
