siRNA Details
Gene Symbol: Tmem102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033433.4 | NP_001028605.2 | 2 | 734 | ||
GenBank mRNA | AK143262.1 | - | 2 | 734 | ||
BC144871.1 | - | 1 | 784 |
Gene Symbol: G630025P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014547.1 | - | 896 | |||
AK036110.1 | - | 768 | ||||
AK137322.1 | - | 1 | 1131 | |||
AK139243.1 | - | 1 | 1121 | |||
AK165723.1 | - | 1 | 1131 |
