siRNA Details
Gene Symbol: Gm1661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033774.2 | NP_001028946.1 | 6 | 846 | ||
NM_001145637.1 | NP_001139109.1 | 6 | 846 | |||
GenBank mRNA | AK132996.1 | - | 6 | 847 | ||
BC147060.1 | - | 4 | 546 | 9056068 | ||
BC147061.1 | - | 4 | 546 | 9056035 | ||
BC171980.1 | - | 4 | 546 |
