siRNA Details
Gene Symbol: Klf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021366.3 | NP_067341.2 | 2 | 6146 | ||
GenBank mRNA | AK079127.1 | - | 1 | 1077 | ||
BC030682.1 | - | 1 | 266 | |||
BC052788.1 | - | 1 | 245 |
