siRNA Details
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160412.1 | NP_001153884.1 | 1 | 162 | ||
GenBank mRNA | AV461889.1 | - | 1 | 172 | ||
EU599038.1 | - | 3 | 252 | |||
EU599039.1 | - | 2 | 236 | |||
EU599040.1 | - | 2 | 236 |
