siRNA Details
Gene Symbol: Dnlz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139504.1 | NP_001132976.1 | 2 | 362 | ||
NM_026828.3 | NP_081104.1 | 2 | 358 | |||
GenBank mRNA | AK004089.1 | - | 2 | 348 | ||
AK142070.1 | - | 2 | 359 | |||
BC139333.1 | - | 2 | 322 | 8862355 | ||
BC139334.1 | - | 2 | 321 | 8862356 | ||
BY266852.1 | - | 2 | 329 |
