siRNA Details
Gene Symbol: Rfk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019437.3 | NP_062310.1 | 4 | 662 | ||
GenBank mRNA | AK002806.1 | - | 4 | 623 | ||
AK008352.1 | - | 4 | 592 | |||
AK010607.1 | - | 4 | 591 | |||
BC033521.1 | - | 4 | 564 | 4527980 | ||
BC051021.1 | - | 4 | 573 | 6315425 |
