siRNA Details
Gene Symbol: Gm17029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035687.1 | - | 408 | |||
AK052342.1 | - | 1372 | ||||
AK084489.1 | - | 1441 |
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019781.2 | NP_062755.1 | 9 | 745 | ||
GenBank mRNA | AF097512.1 | - | 9 | 745 | ||
BC028952.1 | - | 9 | 742 | 5368312 |
