siRNA Details
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019781.2 | NP_062755.1 | 7 | 527 | ||
GenBank mRNA | AF097512.1 | - | 7 | 527 | ||
AK148779.1 | - | 6 | 3454 | |||
BC028952.1 | - | 7 | 524 | 5368312 |
