siRNA Details
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 8 | 1051 | ||
GenBank mRNA | AF116911.1 | - | 8 | 1002 | ||
AK003252.1 | - | 8 | 965 | |||
AK050162.1 | - | 8 | 969 | |||
AK145644.1 | - | 8 | 972 | |||
AK159761.1 | - | 8 | 979 | |||
AK166772.1 | - | 8 | 966 | |||
AK167058.1 | - | 8 | 965 | |||
AK177231.1 | - | 4 | 384 | |||
AK183280.1 | - | 1 | 45 | |||
AX775905.1 | - | 8 | 986 | |||
BC002164.1 | - | 2 | 230 | |||
BC014732.1 | - | 8 | 955 | 4235248 | ||
BC018379.1 | - | 8 | 973 | 4242583 | ||
BC045145.1 | - | 8 | 989 | 6475115 | ||
BC058273.1 | - | 8 | 966 | 6494379 |
