siRNA Details
Gene Symbol: Trip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170907.1 | NP_001164378.1 | 4 | 722 | ||
NM_019797.4 | NP_062771.2 | 7 | 1538 | |||
GenBank mRNA | AF197574.1 | - | 4 | 597 | ||
AF539614.1 | - | 7 | 1421 | |||
AK039024.1 | - | 4 | 607 | |||
AK146298.1 | - | 4 | 605 | |||
AK150752.1 | - | 4 | 599 | |||
BC021316.1 | - | 4 | 610 | 5027899 |
