siRNA Details
Gene Symbol: Trip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019797.4 | NP_062771.2 | 15 | 2553 | ||
GenBank mRNA | AF197574.1 | - | 12 | 1612 | ||
AF539614.1 | - | 15 | 2436 | |||
AK146298.1 | - | 12 | 1620 | |||
BC021316.1 | - | 12 | 1625 | 5027899 |
