siRNA Details
Gene Symbol: Sec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019951.1 | NP_064335.1 | 3 | 312 | ||
GenBank mRNA | AB025405.1 | - | 3 | 312 | ||
AK007449.1 | - | 3 | 310 | |||
AK169560.1 | - | 3 | 325 | |||
BC010484.1 | - | 3 | 304 | 3483051 | ||
BC057885.1 | - | 3 | 504 |
