siRNA Details
Gene Symbol: Prss21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020487.4 | NP_065233.2 | 1, 2 | 136 | ||
GenBank mRNA | AB033822.1 | - | 1, 2 | 122 | ||
AB059415.1 | - | 1, 2 | 134 | |||
AF176209.2 | - | 1, 2 | 136 | |||
AK006271.1 | - | 1, 2 | 108 | |||
AY005145.1 | - | 1, 2 | 113 | |||
BC049588.1 | - | 1, 2 | 150 | |||
BC116755.1 | - | 1, 2 | 134 | 40126074 | ||
BC116757.1 | - | 1, 2 | 134 | 40126076 |
