siRNA Details
Gene Symbol: Adamdec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021475.2 | NP_067450.1 | 1, 2 | 104 | ||
GenBank mRNA | AJ242912.1 | - | 1, 2 | 104 | ||
AK156586.1 | - | 1, 2 | 119 | |||
BC026542.1 | - | 1, 2 | 127 | |||
BC046324.1 | - | 1, 2 | 95 | 6394673 |
