siRNA Details
Gene Symbol: Adamdec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021475.2 | NP_067450.1 | 2 | 156 | ||
GenBank mRNA | AJ242912.1 | - | 2 | 156 | ||
AK156586.1 | - | 2 | 171 | |||
BC026542.1 | - | 2 | 179 | |||
BC046324.1 | - | 2 | 147 | 6394673 |
