siRNA Details
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271843.1 | NP_001258772.1 | 5 | 722 | ||
NM_022026.3 | NP_071309.1 | 5 | 722 | |||
GenBank mRNA | AB037180.1 | - | 5 | 519 | ||
AK050042.1 | - | 5 | 722 | |||
AK154151.1 | - | 5 | 726 | |||
BC024105.1 | - | 6 | 843 | 5096952 | ||
CT010174.1 | - | 5 | 516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK050325.1 | - | 1778 | Chr. - - [-] on Build |
